Beskrivning
- Proteomic discovery tool
- Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
- Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
- Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
- Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
- Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
- Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Rekommenderad förvaring
Store at room temperature
Specifikationer
Specifikationer
| För användning med (applikation) | Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors |
| Kvantitet | 1 Membran |
| Material | PVDF |
| Produktlinje | Novus |