Läs mer
Intedanib, TRC
Produktkod. 30404953
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30404953 100 mg
1 options
Produktkod. 30404953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI666650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nintedanib
CAS 656247-17-5
Molekylformel C31 H33 N5 O4
Kvantitet 100 mg
Synonym 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-, methyl ester, (3Z)-, 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-[[[4-[methyl[(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-, methyl ester, (3Z)- (9CI), BIBF 1120, Nintedanib, Ofev, Vargatef
InChI formel InChI=1S/C31H33N5O4/c1-34-15-17-36(18-16-34)20-27(37)35(2)24-12-10-23(11-13-24)32-29(21-7-5-4-6-8-21)28-25-14-9-22(31(39)40-3)19-26(25)33-30(28)38/h4-14,19,32H,15-18,20H2,1-3H3,(H,33,38)/b29-28-
LEDER COC(=O)c1ccc2\C(=C(\Nc3ccc(cc3)N(C)C(=O)CN4CCN(C)CC4)/c5ccccc5)\C(=O)Nc2c1
IUPAC-namn methyl (3Z)-3-[[4-[methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino]anilino]-phenylmethylidene]-2-oxo-1H-indole-6-carboxylate
Molekylvikt (g/mol) 539.62
Formel vikt 539.2533