missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Integrin alpha 4/CD49d Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-77333-0.025mg
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Integrin alpha 4/CD49d Polyclonal specifically detects Integrin alpha 4/CD49d in Human, Mouse, Rat, Insect samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifikationer
| Integrin alpha 4/CD49d | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least two isoforms are known to exist. The immunogen to this antibody is within the Integrin alpha4 recombinant protein fragment and thus will recognize it. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| P13612 | |
| ITGA4 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Integrin alpha 4. The immunogen is located within amino acids 220 - 270 of Integrin alpha 4. Amino Acid Squence: TTNKYKAFLDKQNQV | |
| 0.025 mg | |
| Cancer, Cellular Markers, Cytoskeleton Markers | |
| 3676 | |
| Human, Mouse, Rat, Insect | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering