missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Integrin alpha E/CD103 Antibody (Ber-ACT8), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97564PEATT594
This item is not returnable.
View return policy
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Flow Cytometry.
Specifications
| Integrin alpha E/CD103 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| ITGAE | |
| HTLV-1 induced human T cell line MAPS16 | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody Ber-ACT8 recognizes an extracellular epitope of CD103 (alpha E integrin), a type I transmembrane glycoprotein primarily found on intestinal intraepithelial lymphocytes. HLDA V; WS Code A067 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| Ber-ACT8 | |
| Flow Cytometry | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
| Mouse | |
| Protein A purified | |
| Innate Immunity | |
| 3682.0 | |
| Human, Primate | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction