missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Integrin beta 1/CD29 Antibody (P4C10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NBP2-36561
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Block/Neutralize.
Specifikationer
| Integrin beta 1/CD29 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot reported in scientific literature (PMID 27550917), Flow Cytometry 1:10, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:100, Block/Neutralize | |
| P05556 | |
| ITGB1 | |
| HT1080 cells human fibrosarcoma | |
| Protein G purified | |
| RUO | |
| Primary | |
| Epitope is not trypsin sensitive. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| P4C10 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Mouse | |
| 88 kDa | |
| 0.1 mL | |
| Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Human, Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering