Learn More
Interference Check Standard AB-2(ICS-AB2) Solution, Specpure™, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041330.AE
Additional Details : Weight : 0.00010kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Interference Check Standard AB-2(ICS-AB2) Solution | |
100 mL | |
UN1789 | |
Liquid |
Matrix: 20% HCl | |
MFCD00803369 | |
Specpure™ |
RUO – Research Use Only