Läs mer
Intermedine, TRC
Produktkod. 30004612
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30004612 0.5 mg 0.5mg
30011094 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004612 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI6666600.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 10285-06-0
Molekylformel C15 H25 N O5
Molekylvikt (g/mol) 299.36
Synonym Butanoic acid, 2,3-dihydroxy-2-(1-methylethyl)-, [(1R,7aR)-2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl]methyl ester, (2S,3R)-, Butanoic acid, 2,3-dihydroxy-2-(1-methylethyl)-, (2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl)methyl ester, [1R-[1α,7(2S*,3R*),7aβ]]-, Intermedine (7CI,8CI), (+)-Intermedine, 3'-epi-Lycopsamine
IUPAC-namn [(7R,8R)-7-hydroxy-5,6,7,8-tetrahydro-3H-pyrrolizin-1-yl]methyl (2S)-2-hydroxy-2-[(1R)-1-hydroxyethyl]-3-methylbutanoate
LEDER CC(C)[C@](O)([C@@H](C)O)C(=O)OCC1=CCN2CC[C@@H](O)[C@@H]12
Specifikationer
Kemiskt namn eller material Intermedine
Kvantitet 0.5 mg
InChI formel InChI=1S/C15H25NO5/c1-9(2)15(20,10(3)17)14(19)21-8-11-4-6-16-7-5-12(18)13(11)16/h4,9-10,12-13,17-18,20H,5-8H2,1-3H3/t10-,12-,13-,15+/m1/s1
Formel vikt 299.1733