Läs mer
Invitrogen™ E-Gel™ Double Comb Agarose Gels, 0.8%
Beskrivning
Innehåller
Each standard box of E-Gel Double Comb Gels contains 18 gels and a manual.
- Supplied in a two-row, mini-gel format
- Each row contains 8 sample wells and 1 well for a marker
- The spacing between wells is compatible with eight-channel pipettors for easy, high-throughput loading
- Self-contained, precast agarose gels designed to provide fast, convenient, and easy electrophoresis
- Each E-Gel™ is ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
- For analyzing PCR products, restriction digests, and plasmid preparations
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Beställningsinfo
Shipping Condition: Room Temperature
Specifikationer
Specifikationer
| Innehåll och lagring | Each standard box of E-Gel™ Double Comb Gels contains 18 gels and a manual. Store at room temperature. |
| Gel Procent | 0.8% |
| Gel typ | E-Gel |
| Wells | 2 x 8-well |
| I Gel Dye | Ethidium Bromide |
| Frakt skick | Room Temperature |
| Well Design | Double Comb |
| Produktlinje | E-Gel |
| Produkttyp | Double Comb Agarose Gel |