Learn More
Invitrogen™ Electrocomp™ TOP10F Electrocomp Kit
Identical to TOP10 with the addition of an F' episome
Brand: Invitrogen™ C66524
Additional Details : Weight : 0.12000kg
Description
- Bacterial/Yeast Strain: TOP10F
Cloning, Transformation
Specifications
Contains enough electrocompetent cells for 75 x 40μL rxns or 50 x 20μL rxns | |
30 x 100 μL |