Learn More
Invitrogen™ GeneCatcher™ gDNA Automated Blood Kit
High sensitivity for archived or poorly stored blood samples
Brand: Invitrogen™ CS2111096
Additional Details : Weight : 0.50000kg
Includes: GeneCatcher Magnetic Beads, Protease, Protease Buffer, GeneCatcher Lysis and Wash Buffers, and GeneCatcher Elution Buffer.
Description
The GeneCatcher™ gDNA Blood Kits allow rapid and efficient extraction of genomic DNA (gDNA) from human blood, including archived or poorly stored blood samples. Genomic DNA is extracted from blood samples using the cost-effective, user-friendly magnetic bead based GeneCatcher Technology without the use of centrifugation
- Purity suited to highly reliable downstream processing
- Validated in many applications, including restriction endonuclease digestion
- Second generation multiplex and single tandem repeat (SGM+STR) profiling and quantitative PCR
- Optimized and designed for scalable and automated gDNA isolation from large blood samples using Tecan Freedom EVO liquid handler
- Fully scalable chemistry in batches of 8
- High purity and yield of extracted genomic DNA yields between 300μg - 500μg per 10mL sample
Automated Genomic DNA Purification, Automated Nucleic Acid Purification, DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Blood (Large-Scale, >1mL)
Order Info
Shipping Condition: Room Temperature
Specifications
The GeneCatcher™ gDNA kits include GeneCatcher™ Magnetic Beads, Protease, Protease Buffer, GeneCatcher™ Lysis and Wash Buffers, and GeneCatcher™ Elution Buffer. All components of the GeneCatcher™ gDNA Blood Kits are shipped at room temperature. Upon receipt, store the protease at +4°C and the remaining kit components at room temperature. All components are guaranteed stable for 6 months when properly stored. | |
Blood | |
High-throughput Compatible | |
> 1 ml Blood Samples (Large-Scale) | |
Room Temperature |
Magnetic Bead | |
Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, Forensic Testing | |
48 Preps | |
GeneCatcher™ | |
Genomic DNA |
For Research Use Only. Not for use in diagnostic procedures.