missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ ReadyPouch™ 1% Agarose Gels, 10 Pouches

Produktkod. 15352242
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10-pack
Förpackningsstorlek:
Paket med 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15352242 10-pack Paket med 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15352242 Leverantör Invitrogen™ Leverantörsnummer A25647

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Innehåller

Ten 100mL pouches and 1mL 0.625mg/mL ethidium bromide dropper

Convenient option for preparing agarose gels

Used for analysis and recovery of DNA for routine applications. Packaged in ready-to-microwave pouch that contains right amount of agarose and buffer for 100mL gel. Vent has to be pouched, heated in a microwave oven, thereafter, add stain, and pour.

  • Convenient and easy-to-use
  • Eliminates risk of agarose boil-overs
  • Clear gel for improved visualization
  • Strong gel structure for better handling and less breakage
  • Improved resolution of very large and very small fragments
  • Comes with an easy-to-use stain dropper
  • Separation range: 100 bp to 12 kb

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifikationer

Koncentration 1%
Form Gel
Material Agarose
Kvantitet 10-pack
Produkttyp Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.