Läs mer
Iodide (from NaI) Standard: I- @ 1000 μg/mL in H2O, VHG™
Produktkod. 16358857
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16358857 100 ml
17979004 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16358857 Leverantör VHG Labs Leverantörsnummer VHGII100
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Iodide (from NaI) Standard: I- @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Iodide
CAS 20461-54-5
Metoder Ion Chromatography
Analyt- eller komponentnamn Iodide
Frakt skick Room Temperature
Molekylformel I
Synonym I−
InChI formel InChI=1S/HI/h1H/p-1
LEDER [I-]
IUPAC-namn iodide
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 126.904
Kvantitet 100 ml
Formel vikt 126.904 g/mol