Learn More
Iodoethane, Sigma-Aldrich™
Product Code. 15888450
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888450 500 mL 500mL
15878450 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888450 Supplier Sigma Aldrich Supplier No. 8008960500
 Restricted Item
Beräknad leverans: 19-11-2026
Stabilized with silver for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-03-6
Specifications
Melting Point -108°C
Density 1.93 g/cm3 at 20°C
Boiling Point 69°C to 73°C/1013 hPa
MDL Number MFCD00001091
Quantity 500 mL
Synonym Iodoethane, Ethyl iodide, Iodoethane
Molecular Weight (g/mol) 155.97
Grade Research