Läs mer
Iodomethylsulbactam (Iodomethyl (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-Dioxide), Mikromol™
Beskrivning
Iodomethylsulbactam (Iodomethyl (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-Dioxide)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Iodomethylsulbactam |
| CAS | 76247-39-7 |
| Analyt- eller komponentnamn | Iodomethylsulbactam |
| Frakt skick | Room Temperature |
| Molekylformel | C9 H12 I N O5 S |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, iodomethyl ester, 4,4-dioxide, (2S,5R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, iodomethyl ester, 4,4-dioxide, (2S-cis)-, Iodomethyl 1,1-dioxopenicillanate, Iodomethyl S,S-dioxopenicillanate, Iodomethyl penicillanate 1,1-dioxide, Sulbactam iodomethyl ester |
| InChI formel | InChI=1S/C9H12INO5S/c1-9(2)7(8(13)16-4-10)11-5(12)3-6(11)17(9,14)15/h6-7H,3-4H2,1-2H3/t6-,7+/m1/s1 |
| LEDER | CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(=O)OCI |
| IUPAC-namn | iodomethyl (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Visa mer |