Läs mer
Iodomethylsulbactam (Iodomethyl (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-Dioxide), Mikromol™
Produktkod. 30463451
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30463451 4 x 25 mg
30052640 25 mg
30049695 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30463451 Leverantör Mikromol Leverantörsnummer MM0308.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Iodomethylsulbactam (Iodomethyl (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-Dioxide), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Iodomethylsulbactam
CAS 76247-39-7
Analyt- eller komponentnamn Iodomethylsulbactam
Frakt skick Room Temperature
Molekylformel C9 H12 I N O5 S
Certifikat och efterlevnad ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, iodomethyl ester, 4,4-dioxide, (2S,5R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, iodomethyl ester, 4,4-dioxide, (2S-cis)-, Iodomethyl 1,1-dioxopenicillanate, Iodomethyl S,S-dioxopenicillanate, Iodomethyl penicillanate 1,1-dioxide, Sulbactam iodomethyl ester
InChI formel InChI=1S/C9H12INO5S/c1-9(2)7(8(13)16-4-10)11-5(12)3-6(11)17(9,14)15/h6-7H,3-4H2,1-2H3/t6-,7+/m1/s1
Lösningstyp Neat
LEDER CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(=O)OCI
IUPAC-namn iodomethyl (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molekylvikt (g/mol) 373.16
Kvantitet 4 x 25 mg
Formel vikt 372.9481 g/mol