Läs mer
Iodophenol Blue, TRC
Produktkod. 30417900
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417900 500 mg
30415229 25 mg
30413131 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT296185500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4430-24-4
Molekylformel C14 H14 F N . H Cl
Molekylvikt (g/mol) 251.727
Synonym 3H-2,1-Benzoxathiole, Phenol deriv., 4,4'-(1,1-Dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-diiodophenol], NSC 36792, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-diiodo-phenol S,S-Dioxide
IUPAC-namn 2-[4-(4-fluorophenyl)phenyl]ethanamine;hydrochloride
LEDER Cl.NCCc1ccc(cc1)c2ccc(F)cc2
Specifikationer
Kemiskt namn eller material Iodophenol Blue
Kvantitet 500 mg
InChI formel InChI=1S/C14H14FN.ClH/c15-14-7-5-13(6-8-14)12-3-1-11(2-4-12)9-10-16;/h1-8H,9-10,16H2;1H
Formel vikt 251.088