Läs mer
Iodoquinol, TRC
Produktkod. 17928114
Change view
Kvantitet:
100 mg
250 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17928114 100 mg -
17908424 250 mg -
17977454 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17928114 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI728400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83-73-8
Molekylformel C9 H5 I2 N O
Molekylvikt (g/mol) 396.95
Synonym 5,7-Diiodoquinolin-8-ol, Clioquinol Imp. C (EP), 5,7-Diiodo-8-hydroxyquinoline, 5,7-Diiodo-8-quinolinol, 5,7-Diiodooxine, 8-Hydroxy-5,7-diiodoquinoline, Di-Quinol, Diamoebin, Diiodohydroxyquin, Diiodohydroxyquinoline, Diiodoquin, Diiodoquinol, Dinoleine, Diodohydroxyquin, Diodoquin, Diodoxylin, Direxiode, Disoquin, Dyodin, Embequin, Enterodiamoebin, Enterosept, Floraquin, Iodoquinol, Ioquin, Lanodoxin, Moebiquin, NSC 74939, NSC 8704, Quinadome, Rafamebin, SS 578, Searlequin, Searlewuin, Sebaquin, Stanquinate, Yodoxin, Zoaquin
IUPAC-namn 5,7-diiodoquinolin-8-ol
LEDER Oc1c(I)cc(I)c2cccnc12
Specifikationer
Kemiskt namn eller material Iodoquinol (5,7-Diiodoquinolin-8-ol)
Kvantitet 100 mg
InChI formel InChI=1S/C9H5I2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
Formel vikt 396.846