Läs mer
Iodosulfuron Methyl Ester Sodium, TRC
Produktkod. 30010153
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30010153 25 g
1 options
Produktkod. 30010153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI72000525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Iodosulfuron-methyl sodium
CAS 144550-36-7
Molekylformel C14 H13 I N5 O6 S . Na
Kvantitet 25 g
Synonym Iodosulfuron-methyl-Na, Sodium 4-iodo-2-methyl-6-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate
InChI formel InChI=1S/C14H14IN5O6S.Na/c1-7-16-12(19-14(17-7)26-3)18-13(22)20-27(23,24)10-6-8(15)4-5-9(10)11(21)25-2;/h4-6H,1-3H3,(H2,16,17,18,19,20,22);/q;+1/p-1
LEDER [Na+].COC(=O)c1ccc(I)cc1S(=O)(=O)[N-]C(=O)Nc2nc(C)nc(OC)n2
IUPAC-namn sodium;(5-iodo-2-methoxycarbonylphenyl)sulfonyl-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoyl]azanide
Molekylvikt (g/mol) 529.24
Formel vikt 528.9529