Läs mer
Iodosulfuron, TRC
Produktkod. 30134695
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134695 1 g
30133374 250 mg
17913435 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7360001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 185119-76-0
Molekylformel C13 H12 I N5 O6 S
Molekylvikt (g/mol) 493.23
Synonym 4-Iodo-2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]benzoic acid, Huzar, Iodosulfuron, Triofensulfuron
IUPAC-namn 4-iodo-2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoic acid
LEDER COc1nc(C)nc(NC(=O)NS(=O)(=O)c2cc(I)ccc2C(=O)O)n1
Specifikationer
Kemiskt namn eller material Iodosulfuron
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C13H12IN5O6S/c1-6-15-11(18-13(16-6)25-2)17-12(22)19-26(23,24)9-5-7(14)3-4-8(9)10(20)21/h3-5H,1-2H3,(H,20,21)(H2,15,16,17,18,19,22)
Formel vikt 492.9553
Procent renhet >95%