missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ion Chromatography Standard: Dichloroacetate @ 500 μg/mL in H2O, VHG™
Beskrivning
Ion Chromatography Standard: Dichloroacetate @ 500 μg/mL in H2O
Specifications
Specifications
| Kemiskt namn eller material | Dichloroacetate |
| CAS | 13425-80-4 |
| Metoder | Ion Chromatography |
| Analyt- eller komponentnamn | Dichloroacetate |
| Frakt skick | Room Temperature |
| Molekylformel | C2 H Cl2 O2 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Acetic acid, 2,2-dichloro-, ion(1-), Acetic acid, dichloro-, ion(1-) (8CI,9CI), 2,2-Dichloroacetate, Dichloroacetate anion, Dichloroacetate ion, Dichloroacetate ion(1-) |
| InChI formel | InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/p-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?