Learn More
Ion Chromatography Standard: Strontium @ 100 μg/mL in H2O, VHG™
Product Code. 17987484
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987484 100 mL 100mL
17997384 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17987484 Supplier VHG Labs Supplier No. VHGISR100
 Restricted Item
Beräknad leverans: 11-01-2027
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Ion Chromatography Standard: Strontium @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Strontium
CAS 7440-24-6
Methods Ion Chromatography
Analyte or Component Names Strontium
Shipping Condition Room Temperature
Molecular Formula Sr
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Sr
InChI Formula InChI=1S/Sr
SMILES [Sr]
IUPAC Name strontium
Solvent or Matrix Water
Molecular Weight (g/mol) 87.62
Quantity 100 mL
Formula Weight 87.906 g/mol