Läs mer
IOX4, TRC
Produktkod. 30136183
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136183 25 mg
30004476 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136183 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI73701025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1154097-71-8
Molekylformel C15H16N6O3
Molekylvikt (g/mol) 328.33
Synonym 6-[2,5-Dihydro-5-oxo-4-(1H-1,2,3-triazol-1-yl)-1H-pyrazol-1-yl]-3-pyridinecarboxylic Acid 1,1-Dimethylethyl Ester;
IUPAC-namn tert-butyl 6-(5-oxo-4-(1H-1,2,3-triazol-1-yl)-2,5-dihydro-1H-pyrazol-1-yl)nicotinate
LEDER CC(C)(C)OC(C1=CN=C(N2NC=C(N3C=CN=N3)C2=O)C=C1)=O
Specifikationer
Kemiskt namn eller material tert-butyl 6-[3-oxo-4-(triazol-1-yl)-1H-pyrazol-2-yl]pyridine-3-carboxylate
Kvantitet 25 mg
InChI formel InChI=1S/C15H16N6O3/c1-15(2,3)24-14(23)10-4-5-12(16-8-10)21-13(22)11(9-18-21)20-7-6-17-19-20/h4-9,18H,1-3H3
Formel vikt 328.13