Läs mer
Ioxitalamic Acid, TRC
Produktkod. 17994785
Change view
Kvantitet:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994785 10 mg
17933885 100 mg
17943115 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994785 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI73810010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28179-44-4
Molekylformel C12 H11 I3 N2 O5
Molekylvikt (g/mol) 643.94
Synonym 3-(Acetylamino)-5-[[(2-hydroxyethyl)amino]carbonyl]-2,4,6-triiodo-benzoic acid, Isophthalamic acid, 5-acetamido-N-(2-hydroxyethyl)-2,4,6-triiodo- (8CI), 3-Acetamido-2,4,6-triiodo-(N-β-hydroxyethyl)isophthalic acid monoamide, AG 58107, Ioxitalamate, Ioxitalamic acid, Ioxithalaminic acid
IUPAC-namn 3-acetamido-5-(2-hydroxyethylcarbamoyl)-2,4,6-triiodobenzoic acid
LEDER CC(=O)Nc1c(I)c(C(=O)O)c(I)c(C(=O)NCCO)c1I
Specifikationer
Kemiskt namn eller material Ioxitalamic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C12H11I3N2O5/c1-4(19)17-10-8(14)5(11(20)16-2-3-18)7(13)6(9(10)15)12(21)22/h18H,2-3H2,1H3,(H,16,20)(H,17,19)(H,21,22)
Formel vikt 643.7802