missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ IP6K1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583520
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: DSDGYINLVA YPYVESETVE QDDTTEREQP RRKHSRRSLH RSGSGSDHKE EKASLSLETS ESSQEAKSP Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 96%.
This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.
Specifikationer
| IP6K1 | |
| Polyclonal | |
| Unconjugated | |
| IP6K1 | |
| 1200016D08Rik; ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; AU022776; IHPK1; inositol hexakisphosphate kinase 1; inositol hexakisphosphate kinase 6; inositol hexaphosphate kinase 1; InsP6; InsP6 kinase 1; InsP6k1; IP6K1; Itpk6; KIAA0263; Pi uptake stimulator; PiUS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92551 | |
| IP6K1 | |
| Recombinant protein corresponding to Human IP6K1. Recombinant protein control fragment (Product #RP-96448). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering