missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP6K1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12057-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.Specifications
IP6K1 | |
Polyclonal | |
Unconjugated | |
IP6K1 | |
IHPK1, InsP6 kinase 1, IP6K1, KIAA0263, PiUS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27399, 50560, 9807 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PD10, Q92551, Q9ESM0 | |
Ip6k1 | |
IP6K1 Fusion Protein Ag2687 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |