Läs mer
Memmert™ IPP Refrigerated Incubator with Peltier Cooling and Twin Display, 256 L, Stainless Steel
Produktkod. 13450229 Handla allt Memmert Produkter
Spänning:
115/230 V
Förpackningsstorlek:
Varje
Produktkod. 13450229
Brand: Memmert™ IPP260plus
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Temperature Range: 0°C to 70°C
  • TwinDISPLAY. Adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-color displays
  • Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
  • Process time does not start until the set temperature is reached
  • German, English, Spanish, French, Polish, Czech, Hungarian language settings
  • Three freely selectable temperature values
  • Temperature (Celsius or Fahrenheit), program time, time zones, summertime/wintertime
  • PRogram stored in case of power failure
  • AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
  • Visual and acoustic alarm
Specifikationer
Certifikat och efterlevnad CE
Kapacitet (metrisk) 256 L
Kammarmaterial Stainless Steel
Material Textured Stainless Steel, Rear Zinc-plated Steel
Pluggtyp German Type
Temperaturområde (metriskt) 0°C to 70°C
Typ Refrigerated Incubator
Visa High-definition TFT-color double display with touchscreen
Inkluderar Works calibration certificate for 10°C and 37°C
Antal dörrar 2
Temperaturnoggrannhet 0.1°C
Mått (L x B x H) 824 W x 1183 H mm
Bredd (metrisk) 824 mm
Bredd (metrisk) Interiör 640 mm
Höjd (metrisk) 1183 mm
Höjd (metrisk) interiör 800 mm
Djup (metrisk) interiör 500 mm
Fraktvikt (metrisk) 164 kg
Vikt (metrisk) 113 kg
Elektriska krav 115/230 V, 50/60 Hz, 820 w
Spänning 115/230 V
Frekvens 50/60 Hz