Läs mer
Ipratropium bromide, MedChemExpress
Produktkod. 16458186
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16458186 100 mg
1 options
Produktkod. 16458186 Leverantör Medchem Express Leverantörsnummer HYB0241
MedChemExpress Ipratropium bromide (Sch 1000) is a muscarinic receptor antagonist, with IC50s of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. Ipratropium bromide relaxes smooth muscle, can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma.
Ipratropium bromide (Sch 1000) is a muscarinic receptor antagonist, with IC50s of 2.9 nM, 2 nM, and 1.7 nM for M1, M2, and M3 receptors, respectively. Ipratropium bromide relaxes smooth muscle, can be used in the research for COPD (chronic obstructive pulmonary disease) and asthma.
Specifikationer
Kemiskt namn eller material Ipratropium bromide
Fysisk form Solid
CAS 22254-24-6
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H30BrNO3
För användning med (applikation) Neuroscience-Neuromodulation
Synonym Sch 1000
Löslighetsinformation H2O : 100 mg/mL (242.51 mM; Need ultrasonic) ∣DMSO : ≥ 35 mg/mL (84.88 mM)
LEDER C[N+]1([C@H]2CC[C@H]1CC(OC(C(CO)C3=CC=CC=C3)=O)C2)C(C)C.[Br-]
Molekylvikt (g/mol) 412.36
Formel vikt 412.36
Procent renhet 98.0%