Learn More
Invitrogen™ iPrep™ Card: gDNA Blood
Descripción
The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.
The following iPrep™ Protocol Cards are available from Invitrogen:
- iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
- iPrep™ Tissue Card for use with the iPrep ChargeSwitch™ Tissue Kit
- iPrep™ Blood Card for use with the iPrep PureLink Blood kit
Especificaciones
Especificaciones
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | iPrep™ |
| Product Line | iPrep |
| Quantity | 1 Card |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.