missing translation for 'onlineSavingsMsg'
Läs mer

Iproplatin, TRC

Produktkod. 30010914
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010914 50 mg
30006562 100 mg
30008887 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30010914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI74052050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 62928-11-4
Molekylformel 2[C3 H9 N] . 2 [Cl] . Pt . 2[O H]
Molekylvikt (g/mol) 418.221
Synonym Dichlorodihydroxybis(isopropylamine)platinum
IUPAC-namn platinum;propan-2-amine;dichloride;dihydroxide
LEDER [OH-].[OH-].[Cl-].[Cl-].[Pt].CC(C)N.CC(C)N

Specifications

Kemiskt namn eller material Iproplatin
Kvantitet 50 mg
InChI formel InChI=1S/2C3H9N.2ClH.2H2O.Pt/c2*1-3(2)4;;;;;/h2*3H,4H2,1-2H3;2*1H;2*1H2;/p-4
Formel vikt 417.0572
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.