Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Iprovalicarb |
| Förpackning | Glass Bottle |
| CAS | 140923-17-7 |
| Färg | White |
| Smältpunkt | 164°C |
| Kokpunkt | Decomposes below the boiling point. |
| Synonym | Isopropyl [(1S)-2-methyl-1-{[(1RS)-1-p-tolylethyl]carbamoyl}propyl]carbamate |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 320.43 |
| Kvantitet | 100 mg |
| Visa mer |