Läs mer
Iprovalicarb, TRC
Produktkod. 30407441
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407441 500 mg
30402933 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30407441 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI747500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Iprovalicarb
CAS 140923-17-7
Molekylformel C18 H28 N2 O3
Kvantitet 500 mg
Synonym 1-Methylethyl N-[(1S)-2-methyl-1-[[[1-(4-methylphenyl)ethyl]amino]carbonyl]propyl]carbamate (ACI), Carbamic acid, [(1S)-2-methyl-1-[[[1-(4-methylphenyl)ethyl]amino]carbonyl]propyl]-, 1-methylethyl ester (9CI), Fencaramid, Iprovalicarb, SZX 0722
InChI formel InChI=1S/C18H28N2O3/c1-11(2)16(20-18(22)23-12(3)4)17(21)19-14(6)15-9-7-13(5)8-10-15/h7-12,14,16H,1-6H3,(H,19,21)(H,20,22)/t14?,16-/m0/s1
LEDER CC(C)OC(=O)N[C@@H](C(C)C)C(=O)NC(C)c1ccc(C)cc1
IUPAC-namn propan-2-yl N-[(2S)-3-methyl-1-[1-(4-methylphenyl)ethylamino]-1-oxobutan-2-yl]carbamate
Molekylvikt (g/mol) 320.43
Formel vikt 320.21