missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IQCH (Human) Recombinant Protein
Human IQCH full-length ORF ( NP_001026885.1, 1 a.a. - 1027 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00064799-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAQNTENHDPVGSILIQIHEDLYQLKEKLTKFSPEEKGETLDIQSLETAIKRTEVGLRIHIEKYLNVVNQNVLTTSVNDESLYTPQASKWLLPTVIDQKSFIFPQESEGTFWQPQRQHSSSLPVFPRAKIKVSKLIKGSNISSLTVLPSSHCTDPYFTPIPVLQADAHKGILSMIERGLIPPTARITFQNPPITPRAAPLHSFDEARKIPTVATFTIPREPPPSPAEVKFFPKKQRSKGKSRRSRGHHDRKAMKVKTPLRALKSLWDYDFLIYDGVIDNTAPDFLAFKEHFSLAWGGIFSLLEHVEKFLRNYAIPEVKIKGNNLVALLPEFELTNKLTRYDLLSVLEDPAHVQMLINLPGQRYKGQDGNSEAAMKIQATWKCYKARKFFLFYRQQKWASGVIAIAWLLYCHKTRLKKILKESRQRHLENFRIRAKHLAANWNRIRTSRRTIIHIPSLGYSQPVREHIADFNTQQNMQLGRLCDILDANVNVIYICSHHMNDELVLYYKKILSLHAAVKSGNLEDRSDLQDRFKIITPEAVNIFPKHHMCLATHLMYSPKAIKRIKNLIRGTEAYIVSGLLHRDDLAVADMLDIPILGSEPELAHLYSTKSGGKRVFDSANVAVPPGIYDIYSQQQMIEQLSQLITDHLQIQRWLFKMDSEFRGNGTAFCDIPSYLKCYKWVLKESSRYGLEDWRKKWAQEPALVKISEELAGILAQHAQPVNEKRFPTWRKFLQTFLSQGGVIEAFPPADNVTNLTVDMLIEPNGKISVLSTGDQLHAESPFISSGTTVPQTSVDPQVLTYLCLQIGKACRMRDVVGYFSIDLVTFIDPSTLEQQVWATGLNLAYSDQLALTQLTLYLTNGHLDCSLSTLEVPRFVPKERKKTKCMSALSMPMLATSRYAVMTTQLRHSNLSLVFHYVFLQICRAHGIGYDLEERQGTVFILYEHLKRHKLGMLTIGEDLQGVLMTFARHLFIIHQEISAPNMQGETNFKTTIADIETILRVTKENKMRFEEEQQSKDDKNLSKPKK
Specifications
NP_001026885.1 | |
IQ motif containing H | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IQCH | |
GST |
64799 | |
Wheat germ expression system | |
10 μg | |
DKFZp434F2114, FLJ12476, NYDSP5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |