missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ IRAK-M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586500
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Purity is > 95% by SDS-PAGE.
IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
Specifikationer
| IRAK-M | |
| Polyclonal | |
| Unconjugated | |
| IRAK3 | |
| 4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y616 | |
| IRAK3 | |
| Synthetic peptide, corresponding to amino acids 500-540 of Human IRAK-M. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering