Läs mer
Tocris Bioscience™ IRAK1/4 Inhibitor I
Produktkod. 18740489
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18740489 50 mg
18768148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18740489 Leverantör Tocris Bioscience™ Leverantörsnummer 5665/50
IRAK4 and IRAK1 inhibitor
IRAK1/4 Inhibitor I is an IRAK4 and IRAK1 inhibitor (IC50 values are 0.2 and 0.3 μM, respectively).
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[1-[2-(4-Morpholinyl)ethyl]-1H-benzimidazol-2-yl]-3-nitrobenzamide
CAS 509093-47-4
Kvantitet 50 mg
Mål Other Kinase Inhibitors
Molekylformel C20H21N5O4
Formel vikt 395.41
Renhet >98%