missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK1, GST Tag Recombinant Protein
IRAK1 Recombinant Human Protein
Marke: Invitrogen™ PR8140B
Weitere Details : Weight : 0.01000kg
Beschreibung
IL-1 receptor associated kinases (IRAKs) involved in signaling via Toll/IL-1 pathways.
Spezifikation
NP_001560 | |
83.4 kDa | |
IRAK1, IRAK, pelle | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
IRAK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.