missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18221-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Interleukin-1 (IL-1) and lipopolysaccharide (LPS) induces cellular responses through IL-1 receptor (IL-1R) and Toll-like receptors (TLR). IL-1R-associated kinases (IRAK, IRAK2, and IRAK-M) regulate the activation of NF-kappa-B and MAP kinase (MAPK) by IL-1R/TLR. A novel member in the IRAK/Pelle family was recently identified and designated IRAK-4. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.Specifications
IRAK4 | |
Polyclonal | |
Unconjugated | |
IRAK4 | |
IPD1, IRAK 4, IRAK4, NY REN 64, REN64 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
266632, 300177, 51135 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R4K2, Q9NWZ3 | |
IRAK4 | |
IRAK4 Fusion Protein Ag12982 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |