missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK4, His Tag Recombinant Protein
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases.
Brand: Invitrogen™ PV4002
Additional Details : Weight : 0.00900kg
Description
IRAKs are involved in signaling via Toll/IL-1 pathways.
Specifications
NP_057207 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
IRAK4 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
55.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
IRAK Kinase Family | |
Recombinant | |
Baculovirus |