missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Irbesartan-d6 (benzyl-d6), CDN
Specifications
Specifications
| Kemiskt namn eller material | Irbesartan-d6 |
| CAS | 2375621-21-7 |
| Molekylformel | C25 D6 H22 N6 O |
| Kvantitet | 0.005 g |
| Synonym | Irbesartan D6 (Benzyl D6), Irbesartan-d6, 2-Butyl-3-[[2'-(1 H-tetrazol-5-yl)biphenyl-4-yl-2,3,5,6-d4]methyl-d2]-1,3-diazaspiro[4.4 ]non-1-en-4-one, 2-Butyl-3-[[2'-(2 H-tetrazol-5-yl)[1,1'-biphenyl-2,3,5,6-d4]-4-yl]methyl-d2]-1,3-diazaspiro[4.4 ]non-1-en-4-one |
| InChI formel | InChI=1 S/C25H28N6O/c1-2-3-10-22-26-25(15-6-7-16-25)24(32)31(22)17-18-11-13-19(14-12-18)20-8-4-5-9-21(20)23-27-29-30-28-23/h4-5,8-9,11-14 H,2-3,6-7,10,15-17H2,1H3,(H,27,28,29,30)/i11D,12 D,13 D,14 D,17D2 |
| LEDER | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1c2ccccc2c3nnn[nH]3)C([2 H])([2 H])N4C(=NC5(CCCC5)C4=O)CCCC |
| IUPAC-namn | 2-butyl-3-[dideuterio-[2,3,5,6-tetradeuterio-4-[2-(1 H-tetrazol-5-yl)phenyl]phenyl]methyl]-1,3-diazaspiro[4.4 ]non-1-en-4-one |
| Molekylvikt (g/mol) | 434.5664 |
| Formel vikt | 434.2701 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?