missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Monoclonal Antibody (3E4), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1844.00 SEK - 4465.00 SEK
Specifications
Antigen | IRF4 |
---|---|
Clone | 3E4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16696444
|
Affymetrix eBioscience
56-9858-80 |
25 μg |
1844.00 SEK
Pack of 2 |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16606454
|
Affymetrix eBioscience
56-9858-82 |
100 μg |
4465.00 SEK
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
IRF4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
IRF4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IRF4 |
3E4 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Rat | |
Human, Mouse | |
Q15306, Q64287 | |
16364, 3662 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |