missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Monoclonal Antibody (3E4), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 56-9858-82
Additional Details : Weight : 0.01000kg
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
IRF4 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q15306, Q64287 | |
IRF4 | |
Affinity chromatography | |
RUO | |
16364, 3662 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
3E4 | |
Alexa Fluor 700 | |
IRF4 | |
AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
Rat | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |