missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ IRF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519504
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Not suitable for use with Immunohistochemistry (Paraffin). This antibody is predicted to react with cow based on sequence homology.
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
Specifikationer
| IRF5 | |
| Polyclonal | |
| Unconjugated | |
| IRF5 | |
| AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27056, 296953, 3663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| P56477, Q13568 | |
| IRF5 | |
| Synthetic peptide conjugated to BSA derived from within residues 450 to the C-terminus of Human IRF5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering