Läs mer
Irganox 1141 (>85%), TRC
Produktkod. 30007229
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007229 50 mg
30008517 10 mg
30005220 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI10121550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134701-20-5
Molekylformel C24H42O
Molekylvikt (g/mol) 346.59
Synonym 2-(Hexadecan-2-yl)-4,6-dimethylphenol;2,4-Dimethyl-6-(1-methylpentadecyl)phenol
IUPAC-namn 2-hexadecan-2-yl-4,6-dimethylphenol
LEDER CCCCCCCCCCCCCCC(C)c1cc(C)cc(C)c1O
Specifikationer
Kemiskt namn eller material Irganox 1141
Kvantitet 50 mg
InChI formel InChI=1S/C24H42O/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-21(3)23-19-20(2)18-22(4)24(23)25/h18-19,21,25H,5-17H2,1-4H3
Formel vikt 346.3236