Learn More
Irgarol 1051 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Product Code. 30318866
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30318866 1 mL 1mL
1 options
Product Code. 30318866 Supplier Dr. Ehrenstorfer Supplier No. DREA14374000CY100
 Restricted Item
Beräknad leverans: 11-01-2027
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Irgarol 1051
CAS 28159-98-0
Analyte or Component Names Irgarol 1051
Shipping Condition Room Temperature
Molecular Formula C11 H19 N5 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C11H19N5S/c1-11(2,3)16-9-13-8(12-7-5-6-7)14-10(15-9)17-4/h7H,5-6H2,1-4H3,(H2,12,13,14,15,16)
SMILES CSc1nc(NC2CC2)nc(NC(C)(C)C)n1
IUPAC Name 4-N-tert-butyl-2-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
Solvent or Matrix Cyclohexane
Molecular Weight (g/mol) 253.37
Quantity 1 mL
Formula Weight 253.1361 g/mol