Läs mer
Irinotecan Carboxylate Sodium Salt, TRC
Produktkod. 30408016
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30408016 5 mg
1 options
Produktkod. 30408016 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI7675105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Irinotecan Carboxylate Sodium Salt
CAS 1329502-92-2
Molekylformel C33H39N4O7.Na
Kvantitet 5 mg
Synonym Indolizino[1,2-b]quinoline-7-acetic acid, 2-[([1,4'-bipiperidin]-1'-ylcarbonyl)oxy]-α,12-diethyl-9,11-dihydro-α-hydroxy-8-(hydroxymethyl)-9-oxo-, sodium salt (1:1), (αS)-, Irinotecan Carboxylate Sodium Salt, (alphaS)-2-[([1,4'-Bipiperidin]-1'-ylcarbonyl)oxy]-alpha,12-diethyl-9,11-dihydro-alpha-hydroxy-8-(hydroxymethyl)-9-oxo-indolizino[1,2-b]quinoline-7-acetic Acid Sodium Salt, CPT 11 Carboxylic Acid Sodium Salt
InChI formel InChI=1S/C33H40N4O7.Na/c1-3-22-23-16-21(44-32(42)36-14-10-20(11-15-36)35-12-6-5-7-13-35)8-9-27(23)34-29-24(22)18-37-28(29)17-26(25(19-38)30(37)39)33(43,4-2)31(40)41;/h8-9,16-17,20,38,43H,3-7,10-15,18-19H2,1-2H3,(H,40,41);/q;+1/p-1/t33-;/m0./s1
LEDER [Na+].CCc1c2CN3C(=O)C(=C(C=C3c2nc4ccc(OC(=O)N5CCC(CC5)N6CCCCC6)cc14)[C@@](O)(CC)C(=O)[O-])CO
IUPAC-namn sodium;(2S)-2-[12-ethyl-8-(hydroxymethyl)-9-oxo-2-(4-piperidin-1-ylpiperidine-1-carbonyl)oxy-11H-indolizino[1,2-b]quinolin-7-yl]-2-hydroxybutanoate
Molekylvikt (g/mol) 626.68
Formel vikt 626.2716