missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron, ACS reagent, low in Mg and Mn compounds, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
1 kg
Specifications
Boiling Point | 3000.0°C |
Melting Point | 1535.0°C |
Quantity | 1 kg |
Merck Index | 15,5137 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 55.84 |
Grade | ACS Reagent |
Chemical Name or Material | Iron |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'