missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) methoxide, 98%, Thermo Scientific Chemicals

Catalog Number p-7049770
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7245-21-8
C2H6FeO2
117.913
MFCD00061474
VAPRHKOWFRYFTF-UHFFFAOYSA-N
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40
14598007
iron(2+);methanolate
C[O-].C[O-].[Fe+2]
This item is not returnable. View return policy
7245-21-8
C2H6FeO2
117.913
MFCD00061474
VAPRHKOWFRYFTF-UHFFFAOYSA-N
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40
14598007
iron(2+);methanolate
C[O-].C[O-].[Fe+2]

Iron(II) methoxide, is used in Engineering & Environmental, Catalysis & Synthesis, and In organic syntheses, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Green
Odor Alcohol-like
Linear Formula Fe(OCH3)2
Quantity 2 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 117.92
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Iron(II) methoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) methoxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'