missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride, anhydrous, 98%, Thermo Scientific Chemicals

Catalog Number p-3257020
Click to view available options
Quantity:
10 g
100 g
1 kg
5 kg
25 kg
7705-08-0
Cl3Fe
162.20
MFCD00011005
RBTARNINKXHZNM-UHFFFAOYSA-K
Ferric chloride
iron(3+) trichloride
[Cl-].[Cl-].[Cl-].[Fe+3]
7705-08-0
Cl3Fe
162.20
MFCD00011005
RBTARNINKXHZNM-UHFFFAOYSA-K
Ferric chloride
iron(3+) trichloride
[Cl-].[Cl-].[Cl-].[Fe+3]

It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursor for polyvinyl chloride (PVC), and copper-based metals in printed circuit boards. Iron (III) chloride is used as catalyst for the reaction of ethylene with chlorine, as a leaching agent, and as a drying reagent in certain reactions. It is a mild oxidizing agent and converts copper(I) chloride in to copper(II) chloride. As a moderately strong Lewis acid, it catalyzes chlorination of aromatic compounds and Friedel-Crafts reactions. Along with sodium iodide, it brings about reduction of organic azides to corresponding amines. It is useful in the detection of phenols and phenolic derivatives, gamma-hydroxybutyric acids, and also in Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 316°C
Melting Point 306°C
Color Black
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Solubility Information Freely soluble in methanol,ethanol,ether,and acetone.
Formula Weight 162.21
Odor Odorless
Sensitivity Hygroscopic
Appearance Black crystalline
Density 2.9 g/cm3
Chemical Name or Material Iron(III) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'