missing translation for 'onlineSavingsMsg'
Läs mer

Iron(III) Nitrate Nonahydrate, Supelco™

Produktkod. 15894370
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 g
1 kg
Förpackningsstorlek:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15894370 1 kg 1kg
15884370 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15894370 Leverantör Supelco Leverantörsnummer 1038831000
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8

Specifications

Kemiskt namn eller material Iron(III) Nitrate Nonahydrate
Smältpunkt 47°C
Kvantitet 1 kg
Molekylformel Fe(NO3)3 · 9 H2O
MDL-nummer MFCD00149708
Synonym Iron(III) nitrate nonahydrate, Iron(III) nitrate nonahydrate
Molekylvikt (g/mol) 404
Kvalitet Research
Densitet 1.68 g/cm3 at 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.