Learn More
Alfa Aesar™ Iron, Oil based standard solution, Specpure™, Fe 1000μg/g
Brand:  Alfa Aesar™ 014220.18
Additional Details : Weight : 0.05000kg
Quantity 50g
Product Code. 15486035
  • 753.00 SEK / 50g
EU Stock 1
Add to basket
Specifications
Specifications
Iron Oil based standard solution
Specpure™
MFCD00010999
Fe 1000μg/g
Liquid
50g
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Iron, Oil based standard solution, Specpure, Fe 1000µg/g
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Mineral oil
SDS