Läs mer
Iron Saccharate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Iron Saccharate |
| CAS | 8047-67-4 |
| Molekylformel | [Na2Fe5O8(OH) • 3H2O]n •(C12H22O11)m |
| Kvantitet | 10 g |
| Synonym | Colliron IV, Feojectin, Ferric Hydroxide Sucrose Complex, Ferrivenin, Ferrum Hausmann, Ferrum Vitis, Ferum Hausmann, Fesin, Hippiron, Iron Oxide Saccharated, Iron Sucrose, Iron Sucrose Complex, Iron Sugar, Iron(III) Hydroxide Sucrose Complex, Iviron, Neoferrum, Proferrin, Saccharated Ferric Oxide, Saccharated Iron, Saccharated Iron Oxide, Sucrofer, Venofer, Venoferrum, XI 921 |
| InChI formel | InChI=1S/3C6H10O8.2Fe/c3*7-1(3(9)5(11)12)2(8)4(10)6(13)14;;/h3*1-4,7-10H,(H,11,12)(H,13,14);;/q;;;2*+3/p-6/t3*1-,2-,3-,4+;;/m000../s1 |
| LEDER | [C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[Fe+3].[Fe+3] |
| Molekylvikt (g/mol) | 736.1 |